2-Cyano-4'-methylbiphenyl ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4'-Methyl-[1,1'-biphenyl]-2-carbonitrile

B21273.06 B21273.14 B21273.22
B21273.06EA 30.4 GBP
B21273.06 B21273.14 B21273.22
2-Cyano-4'-methylbiphenyl ≥98%
2-Cyano-4'-methylbiphenyl

Purity: 98+% CAS No.: 114772-53-1 Molecular Formula: C14H11N Formula Weight: 193.26 Melting Point: 49-53° Boiling Point: 320° UN No.: 3077 MDL No.: MFCD00151805

Formula: C₁₄H₁₁N
MW: 193.26 g/mol
Boiling Pt: >320 °C
Melting Pt: 49…53 °C
Density: 1.170
Storage Temperature: Ambient
MDL Number: MFCD00151805
CAS Number: 114772-53-1
EINECS: 422-310-9
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR