2-Amino-5-nitrophenol 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Hydroxy-4-nitroaniline

B20870.14 B20870.22
B20870.14EA 19.6 GBP
B20870.14 B20870.22
2-Amino-5-nitrophenol 96%
2-Amino-5-nitrophenol

Purity: 96% CAS No.: 121-88-0 Molecular Formula: C6H6N2O3 Formula Weight: 154.13 Melting Point: ca 200° dec. Beilstein Reference: 972974

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: ∼ 200 °C (dec.)
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007692
CAS Number: 121-88-0
EINECS: 204-503-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR