Desiccators, vacuum
Supplier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
DWA215 DWA245
AZLODWA215EA 291 GBP
AZLODWA215 AZLODWA245
Desiccators, vacuum
Desiccators Desiccators, Plastic
Transparent polycarbonate lid and base.
  • Durable with high impact resistance
  • Safely withstand high vacuum pressures
  • Ceramic plate allows the use of incandescent crucibles
Delivery information: Supplied wth plate.