Microtube racks, Axygen® AxyRack™
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
R-80-R R-81-G-PC R-81-R-PC R-81-Y-PC RA-48
AXYGR-80-REA 30.7 GBP
AXYGR-80-R AXYGR-81-G-PC AXYGR-81-R-PC AXYGR-81-Y-PC AXYGRA-48
Microtube racks, Axygen® AxyRack™
Racks Tube Racks
Microtube racks are available in three sizes and are designed to accommodate microtubes or screw-cap microtubes.
The 48-place aluminium rack holds 48 screw-cap tubes of all sizes. Unique ribbed-port design allows one-hand operation, supported by non-skid benchtop pads for a firm platform. Long-lasting aluminium construction. Centre support for added strength and stability.
The 80-well and 50-well racks hold eighty or fifty 1,5/2,0 ml micro and screw-cap microtubes, respectively. These racks resist most chemical and organic solvents and are perfect for storing tubes at cryogenicMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable