Horizontal gel electrophoresis units, Axygen®
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HGB-7 HGB-10 HGB-15 HGB-20
AXYGHGB-7EA 401 GBP
AXYGHGB-7 AXYGHGB-10 AXYGHGB-15 AXYGHGB-20
Horizontal gel electrophoresis units, Axygen®
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Axygen® horizontal gel box is a high quality, full-featured unit designed for exceptional durability and efficiency. These gel boxes feature leak-free external gel casting, which allows the user to cast one gel outside of the box while another gel is running inside. Boxes from 7, 10, 15 and 20cm
  • Flexibility – Each unit comes with 2 sizes of gel trays to optimize applications
  • External gel casting to gel box allows for running gels while casting the next one
  • Ease of use – the Axygen contrast enhancer makes loading gels easier
  • Rugged injection molding is
More Product Information