Anti-SH3GL2 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ARP56550_P050
AVIVARP56550_P050EA 353 GBP
AVIVARP56550_P050
Anti-SH3GL2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SH3GL2 (SH3-domain GRB2-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR