Anti-EVX2 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ARP40119_T100
AVIVARP40119_T100EA 307 GBP
AVIVARP40119_T100
Anti-EVX2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EVX2 (even-skipped homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR