Anti-CLDN9 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ARP33617_T100
AVIVARP33617_T100EA 307 GBP
AVIVARP33617_T100
Anti-CLDN9 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CLDN9 (claudin 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR