Sodium dodecyl sulphate (SDS) ≥85.0% USP, NF, BP, Ph. Eur., pure

Supplier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

142363.0416 142363.0914 142363.1211
APPC142363.0416EA 2050 GBP
APPC142363.0416 APPC142363.0914 APPC142363.1211
Sodium dodecyl sulphate (SDS) ≥85.0% USP, NF, BP, Ph. Eur., pure
Sodium dodecyl sulphate (SDS)

Synonyms: Dodecyl Sulphate Sodium Salt, Laurylsulphate Sodium Salt, SDS, Sodium Laurylsulphate

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Minimum assay 85,0%
Identity according to Pharmacopoeias Passes test
Alkalinity Passes test
Residual solvents (Ph.Eur/USP) Passes test
Non-esterified alcohols 4,0%
Sodium Chloride and Sodium Sulfate (USP-NF) 8,0%
Sodium Chloride and Sodium Sulfate (Ph. Eur.) 8,0%
Total alcohols minimum 59,0%
Heavy metals (as Pb) 0,002%
Residual metals ICP: Class 1A (Pt, Pd) 10 ppm
Residual metals ICP: Class 1B (Ir, Rh, Ru, Os) 10 ppm
Residual metals ICP: Class 1C (Mo, Ni, Cr, V) 25 ppm
Residual metals ICP: Class 2 (Cu, Mn) 250 ppm
Residual metals ICP: Class 3 (Fe, Zn) 1.300 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR