Silver nitrate 99,8-100,5 Argentometric titration Ph. Eur., BP, pure

Supplier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

141459.1606 141459.1608 141459.1609
APPC141459.1606EA 181 GBP
APPC141459.1606 APPC141459.1608 APPC141459.1609
Silver nitrate 99,8-100,5 Argentometric titration Ph. Eur., BP, pure
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444 °C
Melting Pt: 212 °C
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1493
ADR: 5.1 II
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99,8 - 100,5%
Identity according to pharmacopoeias Passes test
Appearance Small crystals
Acidity or alkalinity Passes test
Insoluble matter in H₂O 0,005%
Chloride (Cl) 0,001%
Sulphate (SO₄) 0,002%
Residual solvents (Ph.Eur.) Passes test
Foreign salts 0,02%
Al, Pb, Cu and Bi Passes test
Copper (Cu) 0,0005%
Iron (Fe) 0,0005%
Lead (Pb) 0,001%
Colour White
Solubility in water 2,150 g/l at 20 °C
pH 5,4 - 6,4
Melting point 212 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR