2-Allyloxy-1,1,2,2-tetrafluoroethanesulphonyl fluoride 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC9756-1G PC9756-5G
APOSPC9756-1GEA 82.9 GBP
APOSPC9756-1G APOSPC9756-5G
2-Allyloxy-1,1,2,2-tetrafluoroethanesulphonyl fluoride 97%
2-Allyloxy-1,1,2,2-tetrafluoroethanesulphonyl fluoride
Formula: C₅H₅F₅O₃S
Boiling Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD03095419
CAS Number: 73606-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR