2-Pyrazin-2-yl-1-[4-(trifluoromethyl)phenyl]ethanone 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC9740-1G PC9740-5G
APOSPC9740-1GEA 118 GBP
APOSPC9740-1G APOSPC9740-5G
2-Pyrazin-2-yl-1-[4-(trifluoromethyl)phenyl]ethanone 95+%
2-Pyrazin-2-yl-1-[4-(trifluoromethyl)phenyl]ethanone
Formula: C₁₃H₉F₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD04037908
CAS Number: 88283-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR