3-Nitro-4-[3-(trifluoromethyl)phenoxy]benzenecarbaldehyde 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC9467-1G PC9467-500MG PC9467-5G
APOSPC9467-1GEA 199 GBP
APOSPC9467-1G APOSPC9467-500MG APOSPC9467-5G
3-Nitro-4-[3-(trifluoromethyl)phenoxy]benzenecarbaldehyde 97%
3-Nitro-4-[3-(trifluoromethyl)phenoxy]benzenecarbaldehyde
Formula: C₁₄H₈F₃NO₄
Melting Pt: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00138749
CAS Number: 320416-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR