3-Ethoxy-2-fluorophenol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC910220-1G
APOSPC910220-1GEA 457 GBP
APOSPC910220-1G
3-Ethoxy-2-fluorophenol 95%
3-Ethoxy-2-fluorophenol
Formula: C₈H₉FO₂
MW: 156.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24729359
CAS Number: 1191998-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR