tert-Butyl 3-(bromomethyl)-3-fluoroazetidine-1-carboxylate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC908373-1G
APOSPC908373-1GEA 310 GBP
APOSPC908373-1G
tert-Butyl 3-(bromomethyl)-3-fluoroazetidine-1-carboxylate
tert-Butyl 3-(bromomethyl)-3-fluoroazetidine-1-carboxylate
Formula: C₉H₁₅BrFNO₂
MW: 268.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23105996
CAS Number: 1374658-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR