3-Acetyl-4-fluorobenzonitrile 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC908041-1G
APOSPC908041-1GEA 88.1 GBP
APOSPC908041-1G
3-Acetyl-4-fluorobenzonitrile 95%
3-Acetyl-4-fluorobenzonitrile
Formula: C₉H₆FNO
MW: 163.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015615
CAS Number: 267875-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR