[(Oxido)phenyl(trifluoromethyl)-λ-4-sulphanylidene]dimethylammonium tetrafluoroborate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC906499-1G
APOSPC906499-1GEA 520 GBP
APOSPC906499-1G
[(Oxido)phenyl(trifluoromethyl)-λ-4-sulphanylidene]dimethylammonium tetrafluoroborate 98%
[(Oxido)phenyl(trifluoromethyl)-lambda-4-sulphanylidene]dimethylammonium tetrafluoroborate
Formula: C₉H₁₁BF₇NOS
Storage Temperature: Ambient
CAS Number: 1046786-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR