2-Fluoro-6-(methylsulphonyl)toluene 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC904515-1G PC904515-250MG
APOSPC904515-1GEA 531 GBP
APOSPC904515-1G APOSPC904515-250MG
2-Fluoro-6-(methylsulphonyl)toluene 95%
2-Fluoro-6-(methylsulphonyl)toluene
Formula: C₈H₉FO₂S
Storage Temperature: Ambient
MDL Number: MFCD15527256
CAS Number: 828270-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR