2-Hydroxy-4-(trifluoromethyl)benzyl alcohol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-(Hydroxymethyl)-5-(trifluoromethyl)phenol

PC903933-1G
APOSPC903933-1GEA 53.6 GBP
APOSPC903933-1G
2-Hydroxy-4-(trifluoromethyl)benzyl alcohol 95%
2-Hydroxy-4-(trifluoromethyl)benzyl alcohol
Formula: C₈H₇F₃O₂
MW: 192.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203088
CAS Number: 349-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR