4-Ethoxy-2-fluorotoluene 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC900051-1G
APOSPC900051-1GEA 56.7 GBP
APOSPC900051-1G
4-Ethoxy-2-fluorotoluene 98%
4-Ethoxy-2-fluorotoluene
Formula: C₉H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD18914520
CAS Number: 1305322-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR