2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}acetonitrile

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC8431-1G PC8431-250MG
APOSPC8431-1GEA 630 GBP
APOSPC8431-1G APOSPC8431-250MG
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}acetonitrile
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}acetonitrile
Formula: C₈H₄ClF₃N₂S
Melting Pt: 64…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00215247
CAS Number: 306976-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR