4-Amino-2,5-difluorophenol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2,5-Difluoro-4-hydroxyaniline

PC8310-1G
APOSPC8310-1GEA 136 GBP
APOSPC8310-1G
4-Amino-2,5-difluorophenol
4-Amino-2,5-difluorophenol
Formula: C₆H₅F₂NO
MW: 145.107 g/mol
Melting Pt: 125…130 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094492
CAS Number: 120103-19-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR