2-(Trifluoromethyl)anisole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC8230-1G PC8230-250MG PC8230-5G
APOSPC8230-1GEA 63 GBP
APOSPC8230-1G APOSPC8230-250MG APOSPC8230-5G
2-(Trifluoromethyl)anisole
2-(Trifluoromethyl)anisole
Formula: C₈H₇F₃O
MW: 176.14 g/mol
Melting Pt: –14.3…–14.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00142938
CAS Number: 395-48-2
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR