3-Fluoro-4-iodotoluene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-Fluoro-1-iodo-4-methylbenzene, 1-Fluoro-2-iodo-5-methylbenzene

PC8202-1G
APOSPC8202-1GEA 15.8 GBP
APOSPC8202-1G
3-Fluoro-4-iodotoluene
3-Fluoro-4-iodotoluene
Formula: C₇H₆FI
MW: 236.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143162
CAS Number: 452-79-9
EINECS: 610-236-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR