[5-[(4-Chlorobenzyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC8132-1G PC8132-500MG
APOSPC8132-1GEA 482 GBP
APOSPC8132-1G APOSPC8132-500MG
[5-[(4-Chlorobenzyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol 97%
[5-[(4-Chlorobenzyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Formula: C₁₃H₁₂ClF₃N₂OS
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00172667
CAS Number: 318469-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR