3-Chloro-5-(trifluoromethyl)anisole 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC8005-5G PC8005-1G
APOSPC8005-5GEA 124 GBP
APOSPC8005-5G APOSPC8005-1G
3-Chloro-5-(trifluoromethyl)anisole 98%
3-Chloro-5-(trifluoromethyl)anisole
Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115855
CAS Number: 886497-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR