4-Fluoro-o-anisidine hydrochloride 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC7914-1G PC7914-25G PC7914-5G
APOSPC7914-1GEA 15.8 GBP
APOSPC7914-1G APOSPC7914-25G APOSPC7914-5G
4-Fluoro-o-anisidine hydrochloride 98%
4-Fluoro-o-anisidine hydrochloride
Formula: C₇H₉ClFNO
Melting Pt: 201…209 °C
Storage Temperature: Ambient
MDL Number: MFCD09800850
CAS Number: 178671-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR