2,4-Difluoro-3-methylbenzoyl chloride 98+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC7907-1G
APOSPC7907-1GEA 94.9 GBP
APOSPC7907-1G
2,4-Difluoro-3-methylbenzoyl chloride 98+%
2,4-Difluoro-3-methylbenzoyl chloride
Formula: C₈H₅ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD07777157
CAS Number: 112857-70-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR