1-[2-(Trifluoromethyl)phenyl]propanol 98+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC7868-500MG
APOSPC7868-500MGEA 17.9 GBP
APOSPC7868-500MG
1-[2-(Trifluoromethyl)phenyl]propanol 98+%
1-[2-(Trifluoromethyl)phenyl]propanol
Formula: C₁₀H₁₁F₃O
Storage Temperature: Ambient
MDL Number: MFCD08062381
CAS Number: 878572-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR