7-(Trifluoromethyl)-4-quinolinol 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC7767-25G PC7767-5G
APOSPC7767-25GEA 211 GBP
APOSPC7767-25G APOSPC7767-5G
7-(Trifluoromethyl)-4-quinolinol 97%
7-(Trifluoromethyl)-4-quinolinol
Formula: C₁₀H₆F₃NO
Melting Pt: 266…269 °C
Storage Temperature: Ambient
MDL Number: MFCD00006779
CAS Number: 322-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR