3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 3,3,3-Trifluoro-2-hydroxyisobutyric Acid, 3,3,3-Trifluoro-2-hydroxy-2-methylpropanoic acid, 2-Hydroxy-2-(trifluoromethyl)propionic Acid

PC7674-25G PC7674-1G PC7674-5G
APOSPC7674-25GEA 263 GBP
APOSPC7674-25G APOSPC7674-1G APOSPC7674-5G
3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid 97%
3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid
Formula: C₄H₅F₃O₃
Boiling Pt: 189 °C
Melting Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00190646
CAS Number: 374-35-6
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR