1-[4-(Trifluoromethoxy)phenyl]tetrahydro-2H-imidazol-2-one

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC7661-1G PC7661-5G
APOSPC7661-1GEA 15.8 GBP
APOSPC7661-1G APOSPC7661-5G
1-[4-(Trifluoromethoxy)phenyl]tetrahydro-2H-imidazol-2-one
1-[4-(Trifluoromethoxy)phenyl]tetrahydro-2H-imidazol-2-one
Formula: C₁₀H₉F₃N₂O₂
Melting Pt: 145…146 °C
Storage Temperature: Ambient
MDL Number: MFCD08689725
CAS Number: 162748-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR