Dimethyl(methylthio)sulphonium tetrafluoroborate 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: DMTSF

PC7613-1G PC7613-5G
APOSPC7613-1GEA 89.8 GBP
APOSPC7613-1G APOSPC7613-5G
Dimethyl(methylthio)sulphonium tetrafluoroborate 97%
Dimethyl(methylthio)sulphonium tetrafluoroborate
Formula: C₃H₉BF₄S₂
Storage Temperature: Ambient
MDL Number: MFCD00011806
CAS Number: 5799-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR