4-(Trifluoromethyl)benzenesulphonyl chloride 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-(Trifluoromethyl)benzene-1-sulphonyl chloride

PC7510-100G PC7510-25G PC7510-500G
APOSPC7510-100GEA 73.5 GBP
APOSPC7510-100G APOSPC7510-25G APOSPC7510-500G
4-(Trifluoromethyl)benzenesulphonyl chloride 98%
4-(Trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
Boiling Pt: 118…120 °C/18mmHg
Melting Pt: 27…34 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042422
CAS Number: 2991-42-6
UN: 3261
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR