2-Methylsulphonyl-5-trifluoromethylbenzoic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC7471-1G PC7471-250MG PC7471-5G
APOSPC7471-1GEA 105 GBP
APOSPC7471-1G APOSPC7471-250MG APOSPC7471-5G
2-Methylsulphonyl-5-trifluoromethylbenzoic acid
2-Methylsulphonyl-5-trifluoromethylbenzoic acid
Formula: C₉H₇F₃O₄S
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD09264536
CAS Number: 1000339-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR