1,1,1-Trifluoroacetone 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC7180-100G PC7180-25G PC7180-500G PC7180-250G PC7180-1KG
APOSPC7180-100GEA 269 GBP
APOSPC7180-100G APOSPC7180-25G APOSPC7180-500G APOSPC7180-250G APOSPC7180-1KG
1,1,1-Trifluoroacetone 98%
1,1,1-Trifluoroacetone
Formula: C₃H₃F₃O
Boiling Pt: 21…24 °C
Melting Pt: < –78 °C
Density: 1.252 g/cm³ (25 °C)
Flash Pt: –30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000423
CAS Number: 421-50-1
EINECS: 207-005-9
UN: 1993
ADR: 3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR