1-Ethoxy-2,2,2-trifluoroethanol, Technical Grade

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC7120-100G
APOSPC7120-100GEA 0 GBP
APOSPC7120-100G
1-Ethoxy-2,2,2-trifluoroethanol, Technical Grade
1-Ethoxy-2,2,2-trifluoroethanol
Formula: C₄H₇F₃O₂
Boiling Pt: 104…105 °C/745mmHg
Density: 1.221 g/cm³ (25 °C)
Flash Pt: 39 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000441
CAS Number: 433-27-2
EINECS: 207-087-6
UN: 1989
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR