1,1,1-Trichlorotrifluoroacetone 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC7039-25G PC7039-5G
APOSPC7039-25GEA 881 GBP
APOSPC7039-25G APOSPC7039-5G
1,1,1-Trichlorotrifluoroacetone 97%
1,1,1-Trichlorotrifluoroacetone
Formula: Cl₃CC(O)CF₃
Boiling Pt: 86…90 °C
Density: 1.59 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042128
CAS Number: 758-42-9
UN: 2922
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR