N-Ethyl-4-fluoro-2-nitroaniline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC6534-1G
APOSPC6534-1GEA 44.1 GBP
APOSPC6534-1G
N-Ethyl-4-fluoro-2-nitroaniline
N-Ethyl-4-fluoro-2-nitroaniline
Formula: C₈H₉FN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00798367
CAS Number: 774-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR