1,1,1-Trifluoro-3-phenylacetone 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-Phenyl-1,1,1-trifluoropropan-2-one, 1,1,1-Trifluoro-3-phenyl-2-propanone, (3,3,3-Trifluor-2-propanoyl)benzene, 1,1,1-Trifluoro-3-phenylpropan-2-one

PC6229M-1G PC6229M-25G PC6229M-5G
APOSPC6229M-1GEA 31.5 GBP
APOSPC6229M-1G APOSPC6229M-25G APOSPC6229M-5G
1,1,1-Trifluoro-3-phenylacetone 98%
1,1,1-Trifluoro-3-phenylacetone
Formula: C₉H₇F₃O
Boiling Pt: 51…52 °C/2mmHg
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00068171
CAS Number: 350-92-5
UN: 1224
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR