Perfluorooctane 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Octadecafluorooctane

PC6140A-5G
APOSPC6140A-5GEA 15.8 GBP
APOSPC6140A-5G
Perfluorooctane 98%
Perfluorooctane

Solvent with ability to solubilise molecular oxygen e.g.photooxidation of alkene / epoxidation of alkenes.Synth. Commun., 26, 1861 (1996), ibid 27, 447 (1997).

Formula: C₈F₁₈
MW: 438.06 g/mol
Boiling Pt: 99…100 °C
Melting Pt: -25
Density: 1.73 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042083
CAS Number: 307-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR