[3-Bromo-7-(trifluoromethyl)benzothiophen-2-yl]methanol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC57779-1G PC57779-250MG
APOSPC57779-1GEA 188 GBP
APOSPC57779-1G APOSPC57779-250MG
[3-Bromo-7-(trifluoromethyl)benzothiophen-2-yl]methanol
[3-Bromo-7-(trifluoromethyl)benzothiophen-2-yl]methanol
Formula: C₁₀H₆BrF₃OS
MW: 311.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29432909
CAS Number: 2090924-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR