4-Bromo-3-chloro-2-fluorobenzoic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC56867-1G PC56867-5G PC56867-25G
APOSPC56867-1GEA 94.9 GBP
APOSPC56867-1G APOSPC56867-5G APOSPC56867-25G
4-Bromo-3-chloro-2-fluorobenzoic acid 98%
4-Bromo-3-chloro-2-fluorobenzoic acid
Formula: C₇H₃BrClFO₂
MW: 253.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16622238
CAS Number: 194804-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR