Cobalt(III) fluoride 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

PC5678-100G PC5678-25G PC5678-5G
APOSPC5678-100GEA 164 GBP
APOSPC5678-100G APOSPC5678-25G APOSPC5678-5G
Cobalt(III) fluoride 99%
Cobalt (III) fluoride

Hard fluorinating agent. Replaces organic hydrogen with fluorine, J. Appl. Chem., 1952, 127.

Formula: CoF₃
Density: 3.88 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010942
CAS Number: 10026-18-3
EINECS: 233-062-4
UN: 3084
ADR: 8+5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR