2-Amino-4-[4-(trifluoromethoxy)phenyl]-1,3-thiazole 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC5625-1G
APOSPC5625-1GEA 192 GBP
APOSPC5625-1G
2-Amino-4-[4-(trifluoromethoxy)phenyl]-1,3-thiazole 97%
2-Amino-4-[4-(trifluoromethoxy)phenyl]-1,3-thiazole
Formula: C₁₀H₇F₃N₂OS
Storage Temperature: Ambient
MDL Number: MFCD03094299
CAS Number: 436151-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR