Ethyl 4,4,4-trifluoro-3-hydroxy-2-methylene-3-(trifluoromethyl)butanoate 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC56222-250MG
APOSPC56222-250MGEA 743 GBP
APOSPC56222-250MG
Ethyl 4,4,4-trifluoro-3-hydroxy-2-methylene-3-(trifluoromethyl)butanoate 95%
Ethyl 4,4,4-trifluoro-3-hydroxy-2-methylene-3-(trifluoromethyl)butanoate
Formula: C₈H₈F₆O₃
Storage Temperature: Ambient
MDL Number: MFCD28137304
CAS Number: 152251-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR