6-Fluoroquinoline-2-carboxaldehyde 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC56081-250MG
APOSPC56081-250MGEA 42.2 GBP
APOSPC56081-250MG
6-Fluoroquinoline-2-carboxaldehyde 97%
6-Fluoroquinoline-2-carboxaldehyde
Formula: C₁₀H₆FNO
Boiling Pt: 318 °C
Melting Pt: 113…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06751849
CAS Number: 260430-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR