2-Methoxy-6-(trifluoromethyl)benzoic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC5505-10G PC5505-5G PC5505-1G
APOSPC5505-10GEA 318 GBP
APOSPC5505-10G APOSPC5505-5G APOSPC5505-1G
2-Methoxy-6-(trifluoromethyl)benzoic acid 98%
2-Methoxy-6-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD09832319
CAS Number: 119692-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR