2-Bromo-6-fluorobenzal fluoride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-Bromo-2-(difluoromethyl)-3-fluorobenzene

PC53521-1G PC53521-5G
APOSPC53521-1GEA 54.6 GBP
APOSPC53521-1G APOSPC53521-5G
2-Bromo-6-fluorobenzal fluoride
2-Bromo-6-fluorobenzal fluoride
Formula: C₇H₄BrF₃
Storage Temperature: Ambient
MDL Number: MFCD14698533
CAS Number: 1166820-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR