(R)-6-Fluoro-2,3-dihydrobenzofuran-3-amine hydrochloride 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PC535079-100MG PC535079-250MG
APOSPC535079-100MGEA 51.5 GBP
APOSPC535079-100MG APOSPC535079-250MG
(R)-6-Fluoro-2,3-dihydrobenzofuran-3-amine hydrochloride 97%
(R)-6-Fluoro-2,3-dihydrobenzofuran-3-amine hydrochloride
Formula: C₈H₈FNO·ClH
Storage Temperature: Ambient
MDL Number: MFCD24435222
CAS Number: 2102408-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR